Learn More
Abeba™ Crawler ALU 34555 Shoes
Product Code. 15573744
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
15573744 36 1 pair
15523754 37 1 pair
15543754 38 1 pair
15563754 39 1 pair
15573754 40 1 pair
15533764 41 1 pair
15553764 42 1 pair
15563764 43 1 pair
15553824 44 1 pair
15563824 45 1 pair
15573824 46 1 pair
15553844 47 1 pair
15573844 48 1 pair
13 options
Product Code. 15573744
Brand: Abeba™ 34555/36
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
  • CE EN ISO 20345:2011 SB A, E, FO SRC
  • Microfiber
  • White
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3579)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • velcro fastening
  • Scratch-resistant coating in toe area
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB A, E, FO SRC
Color White
Material Microfiber
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 34555
Toe Material Aluminum