missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15170-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody can bind the close sequences genes.
Carnitine acetyltransferase (CRAT) is a key enzyme in the metabolic pathway in mitochondria, peroxisomes and endoplasmic reticulum. CRAT catalyzes the reversible transfer of acyl groups from an acyl-CoA thioester to carnitine and regulates the ratio of acylCoA/CoA in the subcellular compartments. Different subcellular localizations of the CRAT mRNAs are thought to result from alternative splicing of the CRAT gene suggested by the divergent sequences in the 5′ region of peroxisomal and mitochondrial CRAT cDNAs and the location of an intron where the sequences diverge. The alternatively splicing of this gene results in three distinct isoforms, one of which contains an N-terminial mitochondrial transit peptide, and has been shown to be located in mitochondria.Specifications
| CRAT | |
| Polyclonal | |
| Unconjugated | |
| CRAT | |
| CACP, Carnitine acetylase, carnitine acetyltransferase, Carnitine O acetyltransferase, CAT, CAT1, CRAT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12908, 1384, 311849 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P43155, P47934, Q704S8 | |
| Crat | |
| CRAT Fusion Protein Ag7422 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction