missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Craniofacial Development Protein 1 Monoclonal antibody specifically detects Craniofacial Development Protein 1 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Craniofacial Development Protein 1 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 5B7 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_006315 |
| Gene Alias | BCNTBUCENTAUR, Bucentaur, CP27phosphoprotein (Bucentaur), craniofacial development protein 1, p97, SWC5, Yeti |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?