missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRACR2A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15206-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognizes both of CRACR2A-c and CRACR2A-a.
CRACR2A is a Ca(2+)-binding protein that plays a key role in store-operated Ca(2+) entry (SOCE) in T-cells by regulating CRAC channel activation. CRACR2A acts as a cytoplasmic calcium-sensor that facilitates the clustering of ORAI1 and STIM1 at the junctional regions between the plasma membrane and the endoplasmic reticulum upon low Ca(2+) concentration. Further, CRACR2A regulates CRAC channel activation, including translocation and clustering of ORAI1 and STIM1. Upon increase of cytoplasmic Ca(2+) resulting from opening of CRAC channels, CRACR2A dissociates from ORAI1 and STIM1, thereby destabilizing the ORAI1-STIM1 complex.Specifications
| CRACR2A | |
| Polyclonal | |
| Unconjugated | |
| CRACR2A | |
| CRAC channel regulator 2A, CRACR2A, EFCAB4B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84766 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BSW2 | |
| CRACR2A | |
| CRACR2A Fusion Protein Ag7358 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction