missing translation for 'onlineSavingsMsg'
Learn More

CRACR2A Mouse anti-Human, Clone: 4C1F5, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66787-1-IG-150UL

 View more versions of this product

Product Code. 16838985

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody recognizes both of CRACR2A-c and CRACR2A-a.

CRACR2A is a Ca(2+)-binding protein that plays a key role in store-operated Ca(2+) entry (SOCE) in T-cells by regulating CRAC channel activation. CRACR2A acts as a cytoplasmic calcium-sensor that facilitates the clustering of ORAI1 and STIM1 at the junctional regions between the plasma membrane and the endoplasmic reticulum upon low Ca(2+) concentration. Further, CRACR2A regulates CRAC channel activation, including translocation and clustering of ORAI1 and STIM1. Upon increase of cytoplasmic Ca(2+) resulting from opening of CRAC channels, CRACR2A dissociates from ORAI1 and STIM1, thereby destabilizing the ORAI1-STIM1 complex.
Specifications

Specifications

CRACR2A
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BSW2
CRACR2A
CRACR2A Fusion Protein Ag7525
150 μL
Primary
Human
Antibody
IgG1
Western Blot
4C1F5
Unconjugated
CRACR2A
CRAC channel regulator 2A, CRACR2A, DKFZp686G13246, EFCAB4B, FLJ33046, FLJ33805
Mouse
Protein G
RUO
84766
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.