missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRACC/SLAMF7 Antibody (MM0200-6G17), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12206AF594
This item is not returnable.
View return policy
Description
CRACC/SLAMF7 Monoclonal specifically detects CRACC/SLAMF7 in Human samples. It is validated for Flow Cytometry.
Specifications
| CRACC/SLAMF7 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SLAMF7 | |
| Human recombinant CRACC/SLAMF7 | |
| Immunology | |
| 57823 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| MM0200-6G17 | |
| Flow Cytometry | |
| 19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7 | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction