missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRACC Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5148546

Product Code. 30534893

  • £399.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

SLAMF7 contains one Ig-like C2-type (immunoglobulin-like) domain. Isoform 1 mediates NK cell activation through a SAP-independent extracellular signal-regulated ERK-mediated pathway. It may play a role in lymphocyte adhesion. Isoform 3 does not mediate any activation. SAP can bind the cytoplasmic tail of isoform 1 when phosphorylated in the presence of Fyn (in vitro). SLAMF7 is expressed in spleen, lymph node, peripheral blood leukocytes, bone marrow, small intestine, stomach, appendix, lung and trachea. Expression was detected in NK cells, activated B-cells, NK-cell line but not in promyelocytic, B-, or T-cell lines. The isoform 3 is expressed at much lower level than isoform 1. There are three named isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CRACC
Polyclonal
Unconjugated
SLAMF7
19A; 19A24; 19A24 protein; 4930560D03Rik; CD2 subset 1; CD2-like receptor activating cytotoxic cells; CD2-like receptor-activating cytotoxic cells; CD319; CRACC; CS1; Leukocyte cell-surface antigen; membrane protein FOAP-12; Novel Ly9; novel LY9 (lymphocyte antigen 9) like protein; protein 19A; RGD1560190; SLAM family member 7; SLAMF7; UNQ576/PRO1138
Goat
Antigen affinity chromatography
RUO
57823
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q9NQ25
SLAMF7
Mouse myeloma cell line NS0-derived recombinant human CRACC/SLAMF7, Lys27-Ser225.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.