missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRAC intermediate 2, MedChemExpress
Description
CRAC intermediate 2 is a intermediate compound for CRAC inhibitor synthesis, extracted from patent WO 2013059666A1.
Specifications
Specifications
| Chemical Name or Material | CRAC intermediate 2 |
| Physical Form | Solid |
| CAS | 123066-64-8 |
| Color | Light Yellow |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C11H7F6N3 |
| Solubility Information | DMSO : ≥ 52 mg/mL (176.16 mM) |
| SMILES | NC1=CC=C(N2N=C(C(F)(F)F)C=C2C(F)(F)F)C=C1 |
| Molecular Weight (g/mol) | 295.18 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?