missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRAC intermediate 1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
CRAC intermediate 1 is a key intermediate in the chemical synthesis of a series of CRAC channel inhibitors, detailed information can be found in Patent WO 2010122089 A1, intermediate 9.
Specifications
Specifications
| Chemical Name or Material | CRAC intermediate 1 |
| CAS | 1249343-86-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C10H7F2N3O |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 50 mg/mL (224.03 mM) |
| SMILES | O=C(C1=C(F)C=CC=C1F)NC2=NNC=C2 |
| Molecular Weight (g/mol) | 223.18 |
| Formula Weight | 223.18 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction