missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRABP2 Recombinant Rabbit Monoclonal Antibody (24GB4790)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557365
This item is not returnable.
View return policy
Description
CRABP2 Recombinant Monoclonal Antibody for Western Blot, Flow
Cellular retinoic acid binding protein (CRABP) is a low molecular weight protein with unknown function and existing in two isoforms, CRABP1 and CRABP2. The predicted amino acid sequences of human CRABP1 and CRABP2 exhibits 99.3 and 93.5% identity to mouse CRABP1 and CRABP2, respectively. The CRABP2 isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. The CRABP2 gene localized on human chromosome 1q21.3 is transcriptionally regulated by a newly synthesized regulatory protein. The protein is expressed in human adult liver.
Specifications
| CRABP2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CRABP2 | |
| AI893628; cellular lipid-binding protein; cellular retinoic acid binding protein 2; cellular retinoic acid binding protein II; cellular retinoic acid-binding protein 2; Cellular retinoic acid-binding protein II; Crabp2; Crabp-2; CrabpII; CRABP-II; RBP6; RP11-66D17.4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12904, 1382 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4790 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P22935, P29373 | |
| CRABP2 | |
| Recombinant protein of human CRABP2 (aa 1-138). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction