missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRABP2 Rabbit anti-Mouse, Clone: 101, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90627-100UL
This item is not returnable.
View return policy
Description
CRABP2 Monoclonal specifically detects CRABP2 in Mouse samples. It is validated for ELISA.
Specifications
| CRABP2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 101 | |
| ELISA 1:5000-1:10000 | |
| cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6 | |
| Recombinant Mouse CRABP2 protein | |
| 100 μg | |
| Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
| 1382 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction