missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRABP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582140
This item is not returnable.
View return policy
Description
Immunogen sequence: IKQEGDTFYI KTSTTVRTTE INFKVGEEFE EQTVDGRPCK SLVKWESENK MVCEQKLLKG EGPKTSWTRE LTNDGELILT MTADDVVCTR VYVRE Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 95%.
Cellular retinoic acid binding protein (CRABP) is a low molecular weight protein with unknown function and existing in two isoforms, CRABP1 and CRABP2. The predicted amino acid sequences of human CRABP1 and CRABP2 exhibits 99.3 and 93.5% identity to mouse CRABP1 and CRABP2, respectively. The CRABP2 isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. The CRABP2 gene localized on human chromosome 1q21.3 is transcriptionally regulated by a newly synthesized regulatory protein. The protein is expressed in human adult liver.
Specifications
| CRABP2 | |
| Polyclonal | |
| Unconjugated | |
| CRABP2 | |
| AI893628; cellular lipid-binding protein; cellular retinoic acid binding protein 2; cellular retinoic acid binding protein II; cellular retinoic acid-binding protein 2; Cellular retinoic acid-binding protein II; Crabp2; Crabp-2; CrabpII; CRABP-II; RBP6; RP11-66D17.4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1382 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P29373 | |
| CRABP2 | |
| Recombinant protein corresponding to Human CRABP2. Recombinant protein control fragment (Product #RP-96785). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering