missing translation for 'onlineSavingsMsg'
Learn More

CRABP2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A5F3, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66468-1-IG-150UL

 View more versions of this product

Product Code. 16818375

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cellular retinoic acid binding protein (CRABP) is a low molecular weight protein with unknown function and existing in two isoforms, CRABP1 and CRABP2. The predicted amino acid sequences of human CRABP1 and CRABP2 exhibits 99.3 and 93.5% identity to mouse CRABP1 and CRABP2, respectively. The CRABP2 isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. The CRABP2 gene localized on human chromosome 1q21.3 is transcriptionally regulated by a newly synthesized regulatory protein. The protein is expressed in human adult liver.
Specifications

Specifications

CRABP2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P22935, P29373, P51673
CRABP2
CRABP2 Fusion Protein Ag0309
150 μL
Primary
Human, Mouse, Pig, Rat
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
1A5F3
Unconjugated
CRABP2
CRABP II, CRABP2, RBP6
Mouse
Protein G
RUO
100155151, 12904, 1382, 29563
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.