missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRABP2 Monoclonal Antibody (5CRA-3B3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA3017
This item is not returnable.
View return policy
Description
MA3-017 has been used successfully in Western blotting and Immunofluorescence. This antibody was referenced in publications to perform in IHC.
Cellular retinoic acid binding protein (CRABP) is a low molecular weight protein with unknown function and existing in two isoforms, CRABP1 and CRABP2. The predicted amino acid sequences of human CRABP1 and CRABP2 exhibits 99.3 and 93.5% identity to mouse CRABP1 and CRABP2, respectively. The CRABP2 isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. The CRABP2 gene localized on human chromosome 1q21.3 is transcriptionally regulated by a newly synthesized regulatory protein. The protein is expressed in human adult liver.
Specifications
| CRABP2 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.05% sodium azide | |
| P29373 | |
| CRABP2 | |
| recombinant protein corresponding to full length human CRABPII. | |
| RUO | |
| 1382 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 5CRA-3B3 | |
| Unconjugated | |
| CRABP2 | |
| AI893628; cellular lipid-binding protein; cellular retinoic acid binding protein 2; cellular retinoic acid binding protein II; cellular retinoic acid-binding protein 2; Cellular retinoic acid-binding protein II; Crabp2; Crabp-2; CrabpII; CRABP-II; RBP6; RP11-66D17.4 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction