missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CRABP2 Polyclonal antibody specifically detects CRABP2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
Specifications
| Antigen | CRABP2 |
| Applications | Immunoprecipitation |
| Conjugate | mFluor Violet 610 SE |
| Gene Alias | cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6 |
| Regulatory Status | RUO |
| Research Discipline | Core ESC Like Genes, Neuroscience, Stem Cell Markers |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?