missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRABP2 Antibody (JE65-66), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32214
This item is not returnable.
View return policy
Description
CRABP2 Monoclonal antibody specifically detects CRABP2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| CRABP2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:400 | |
| cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6 | |
| Recombinant protein within human CRABP2 aa 1-138/138. (Uniprot: P29373) | |
| 100 μL | |
| Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
| 1382 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| JE65-66 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction