missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRABP2 Antibody (101), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90627V
This item is not returnable.
View return policy
Description
CRABP2 Monoclonal specifically detects CRABP2 in Mouse samples. It is validated for ELISA.
Specifications
| CRABP2 | |
| Monoclonal | |
| DyLight 405 | |
| cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CRABP2 (P22935; Pro 2-Glu 138). | |
| 0.1 mL | |
| Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
| 1382 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 101 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction