missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CRABP2 Monoclonal specifically detects CRABP2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | CRABP2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 16 |
| Conjugate | Alexa Fluor 750 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human CRABP2 (NP_001869.1; Pro2-Glu138). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?