missing translation for 'onlineSavingsMsg'
Learn More

CRABP2 Antibody (004), DyLight 594, Novus Biologicals™

Product Code. 18340241 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340241 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340241 Supplier Novus Biologicals Supplier No. NBP289836DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CRABP2 Monoclonal specifically detects CRABP2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRABP2
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 4
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CRABP2 (NP_001869.1; Pro2-Glu138).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1382
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.