missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRABP1 Recombinant Rabbit Monoclonal Antibody (23GB5105), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | CRABP1 |
|---|---|
| Clone | 23GB5105 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089150
![]() |
Invitrogen™
MA552792 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087480
|
Invitrogen™
MA552793 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CRABP1 Recombinant Monoclonal antibody specifically detects CRABP1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CRABP1 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P29762, P62965 | |
| 12903, 1381 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5105 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI326249; cCRABP-I; cellular retinoic acid binding protein 1; cellular retinoic acid binding protein I; cellular retinoic acid binding protein I (CRABP-I) homologue; cellular retinoic acid-binding protein 1; Cellular retinoic acid-binding protein I; cellular retinoic-acid-binding protein; CRABP; Crabp1; Crabp-1; CrabpI; CRABP-I; RBP5; Rbp-5; retinoic acid-binding protein | |
| A synthesized peptide derived from human CRABP1 (100-137AA). | |
| Primary | |
| -20°C | |
| CRABP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title