missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRABP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103724
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CRABP1.
This gene encodes a specific binding protein for a vitamin A family member and is thought to play an important role in retinoic acid-mediated differentiation and proliferation processes. It is structurally similar to the cellular retinol-binding proteins, but binds only retinoic acid at specific sites within the nucleus, which may contribute to vitamin A-directed differentiation in epithelial tissue.
Specifications
| CRABP1 | |
| Polyclonal | |
| Unconjugated | |
| CRABP1 | |
| AI326249; cCRABP-I; cellular retinoic acid binding protein 1; cellular retinoic acid binding protein I; cellular retinoic acid binding protein I (CRABP-I) homologue; cellular retinoic acid-binding protein 1; Cellular retinoic acid-binding protein I; cellular retinoic-acid-binding protein; CRABP; Crabp1; Crabp-1; CrabpI; CRABP-I; RBP5; Rbp-5; retinoic acid-binding protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12903, 1381, 25061 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P29762, P62965, P62966 | |
| CRABP1 | |
| A synthesized peptide derived from human CRABP1(Accession P29762), corresponding to amino acid residues Q50-L100. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction