missing translation for 'onlineSavingsMsg'
Learn More

CRABP1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£157.00 - £358.00

Specifications

Antigen CRABP1
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18677231
View Documents
Novus Biologicals
NBP2-92084-0.02ml
0.02 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18623380
View Documents
Novus Biologicals
NBP2-92084-0.1ml
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

CRABP1 Polyclonal antibody specifically detects CRABP1 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

CRABP1
Western Blot
Unconjugated
Rabbit
Stem Cell Markers
PBS with 50% glycerol, pH7.3.
1381
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Mouse
cellular retinoic acid binding protein 1, CRABP, CRABPI, CRABP-Icellular retinoic acid-binding protein 1, RBP5Cellular retinoic acid-binding protein I
Recombinant fusion protein containing a sequence corresponding to amino acids 1-137 of human CRABP1 (NP_004369.1). MPNFAGTWKMRSSENFDELLKALGVNAMLRKVAVAAASKPHVEIRQDGDQFYIKTSTTVRTTEINFKVGEGFEEETVDGRKCRSLATWENENKIHCTQTLLEGDGPKTYWTRELANDELILTFGADDVVCTRIYVRE
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.