missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRABP1 Antibody (1A1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-37070CL1
This item is not returnable.
View return policy
Description
CRABP1 Monoclonal antibody specifically detects CRABP1 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| CRABP1 | |
| Monoclonal | |
| CoraFluor 1 | |
| cellular retinoic acid binding protein 1, CRABP, CRABPI, CRABP-Icellular retinoic acid-binding protein 1, RBP5Cellular retinoic acid-binding protein I | |
| Recombinant human CRABP1 (1-137aa) purified from E. coli | |
| 0.1 mL | |
| Stem Cell Markers | |
| 1381 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 1A1 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction