Learn More
CPVL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA588504
Description
Immunogen sequence: YVVTSNMTLR DRDFPWTTTL SMLYIDNPVG TGFSFTDDTH GYAVNEDDVA RDLYSALIQF FQIFPEYKNN DFYVTGESYA GKYVPAIAHL IHSLNPVREV KINLNGIAIG DGYSDPESII GGYAEFLYQI GLLDEKQKKY FQKQCHECIE HIRKQNWFEA FEILDKLLDG DLTSDPSYFQ NVTGCSNYYN FLRCTEPEDQ LYYVKFLSLP EVRQAIHVGN QTFNDGTIVE KYLREDTVQS VKPWLTEIMN NYKVLIYNGQ LDIIVAAALT ERSLMGMDWK GSQEYKKAEK KVWKIFKSDS EVAGYIRQAG DFHQVIIRGG GHILPYDQPL RAFDMINRFI YGKGWDPYVG; Positive Samples: 293T, Rat brain
The protein encoded by this gene is a carboxypeptidase and bears strong sequence similarity to serine carboxypeptidases. Carboxypeptidases are a large class of proteases that act to cleave a single amino acid from the carboxy termini of proteins or peptides. The exact function of this protein, however, has not been determined.
Specifications
| CPVL | |
| Polyclonal | |
| Unconjugated | |
| Cpvl | |
| 4933436L16Rik, carboxypeptidase WUG, carboxypeptidase, vitellogenic like, carboxypeptidase, vitellogenic-like, CP-Mac carboxypeptidase, CPVL, HVLP, probable serine carboxypeptidase CPVL, PSEC0124, tcag7.235, UNQ197/PRO223, VCP-like protein, vitellogenic carboxypeptidase-like protein, VLP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 502774, 54504 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.69 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal, pH 7.3 | |
| Q4QR71, Q9H3G5 | |
| Cpvl | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 127-476 of human CPVL (NP_0619022) | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.