Learn More
Invitrogen™ CPT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109586
Description
Immunogen sequence: ATTDSTVTVQ KLNFELTDAL KTGITAAKEK FDATMKTLTI DCVQFQRGGK EFLKKQKLSP DAVAQLAFQM AFLRQYGQTV ATYESCSTAA FKHGRTETIR PASVYTKRCS EAFVREPSRH SAGELQQMMV ECSKYHGQLT KEAAMGQGFD RHLFALRHLA AAKGIILPEL YLDPAYGQIN HNVLSTSTLS SPAVNLGGFA PVVSDGFGVG YAVHDNWIGC NVSSYPGRNA REFLQCVEKA LEDMFDALEG KSIKS.
Carnitine palmitoyltransferase II precursor (CPT2) is a nuclear protein which is transported to the mitochondrial inner membrane. CPT2 together with carnitine palmitoyltransferase I oxidizes long-chain fatty acids in the mitochondria. Defects in this gene are associated with mitochondrial long-chain fatty-acid (LCFA) oxidation disorders.
Specifications
| CPT2 | |
| Polyclonal | |
| Unconjugated | |
| CPT2 | |
| AI323697; Carnitine O-palmitoyltransferase 2, mitochondrial; carnitine palmitoyltransferase 2; carnitine palmitoyltransferase II; CPT II; CPT1; CPT2; Cpt-2; CPTASE; CPTII; IIAE4; OTTHUMP00000010493; testicular secretory protein Li 13 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12896, 1376, 25413 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P18886, P23786, P52825 | |
| CPT2 | |
| Immunogen is Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.