missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPT1C Recombinant Rabbit Monoclonal Antibody (6N7B2)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | CPT1C |
|---|---|
| Clone | 6N7B2 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284442
|
Invitrogen™
MA558197 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30284212
![]() |
Invitrogen™
MA558198 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: MAEAHQAVGF RPSLTSDGAE VELSAPVLQE IYLSGLRSWK RHLSRFWNDF.
This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein regulates the beta-oxidation and transport of long-chain fatty acids into mitochondria, and may play a role in the regulation of feeding behavior and whole-body energy homeostasis. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.Specifications
| CPT1C | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| F1LN46, Q8BGD5 | |
| 308579, 78070 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-50 of human CPT1C (NP_689572.1). | |
| Antibody |
| 6N7B2 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse, Rat | |
| CPT1C | |
| 6530437J22Rik; 9630004I06Rik; Carnitine O-palmitoyltransferase 1, brain isoform; carnitine O-palmitoyltransferase I, brain isoform; carnitine palmitoyltransferase 1, brain; carnitine palmitoyltransferase 1c; carnitine palmitoyltransferase I related C; CATL1; CPT IC; CPT I-C; CPT1-B; Cpt1c; CPT1P; CPTI-B; CPTIC; FLJ23809; SPG73 | |
| CPT1C | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title