missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPT1C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548206
This item is not returnable.
View return policy
Description
CPT1C Polyclonal Antibody for Western Blot, Flow
This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein regulates the beta-oxidation and transport of long-chain fatty acids into mitochondria, and may play a role in the regulation of feeding behavior and whole-body energy homeostasis. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.
Specifications
| CPT1C | |
| Polyclonal | |
| Unconjugated | |
| CPT1C | |
| 6530437J22Rik; 9630004I06Rik; Carnitine O-palmitoyltransferase 1, brain isoform; carnitine O-palmitoyltransferase I, brain isoform; carnitine palmitoyltransferase 1, brain; carnitine palmitoyltransferase 1c; carnitine palmitoyltransferase I related C; CATL1; CPT IC; CPT I-C; CPT1-B; Cpt1c; CPT1P; CPTI-B; CPTIC; FLJ23809; SPG73 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 126129 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.27 | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8TCG5 | |
| CPT1C | |
| KLH conjugated synthetic peptide between 752-782 amino acids from the C-terminal region of human CPT1C. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction