missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT1C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68811
This item is not returnable.
View return policy
Description
CPT1C Polyclonal antibody specifically detects CPT1C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CPT1C | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| carnitine O-palmitoyltransferase 1, brain isoform, Carnitine O-palmitoyltransferase I, brain isoform, carnitine palmitoyltransferase 1CCPT1-B, carnitine palmitoyltransferase I related C, CATL1, CPT IC, CPT1P, CPTI-B, CPTIC, EC 2.3.1.21, FLJ23809 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AQVRTSLKTQAAEALEAVEGAAFFVSLDAEPAGLTREDPAASLDAYAHALLAG | |
| 100 μg | |
| Cardiovascular Biology, Endocrinology, metabolism, Neuroscience, Signal Transduction | |
| 126129 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction