missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CPT1B Recombinant Rabbit Monoclonal Antibody (0R3Z6)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557928

 View more versions of this product

Product Code. 30284332

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: FNTTRIPGKD TDVLQHLSDS RHVAVYHKGR FFKLWLYEGA RLLKPQDLEM QFQRILDDPS PPQPGEEKLA ALTAGGRVEW AQARQAFFSS GKNKAALEAI ERAAFFVALD EESYSYDPED EASLSLYGKA LLHGNCYNRW FDKSFTLISF KNGQLGLNAE HAWADAPIIG HLWEFVLGTD SFHLGYTE.

The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and read-through transcripts are expressed from the upstream locus that include exons from this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CPT1B
Recombinant Monoclonal
1.43 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Q63704, Q924X2
CPT1B
Recombinant fusion protein containing a sequence corresponding to amino acids 313-500 of human CPT1B (NP_004368.1).
100 μL
Primary
Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot
0R3Z6
Unconjugated
CPT1B
carnitine O-palmitoyltransferase 1, muscle isoform; carnitine O-palmitoyltransferase 1, muscle isoform; synaptonemal complex central element protein 3; carnitine O-palmitoyltransferase 1B; carnitine O-palmitoyltransferase I, mitochondrial muscle isoform; carnitine O-palmitoyltransferase I, muscle isoform; Carnitine palmitoyltransferase 1 beta, muscle isoform; Carnitine palmitoyltransferase 1, muscle; carnitine palmitoyltransferase 1A like; carnitine palmitoyltransferase 1B; carnitine palmitoyltransferase 1B (muscle); carnitine palmitoyltransferase 1B isoform a; carnitine palmitoyltransferase 1b, muscle; carnitine palmitoyltransferase I; carnitine palmitoyltransferase Ibeta 1; carnitine palmitoyltransferase Ibeta 2; carnitine palmitoyltransferase Ibeta 3; carnitine palmitoyltransferase I-like protein; CPT I; Cpt1; cpt1al; CPT1B; CPT1M; Cpt1-m; CPTI; CPTIB; CPT-IB; CPT-Ibeta 1; CPT-Ibeta 3; CPTI-M; EC 2.3.1.21; FLJ55729; FLJ58750; hypothetical protein LOC449677; KIAA1670; LOW QUALITY PROTEIN: carnitine O-palmitoyltransferase 1, muscle isoform; MCCPT1; MCPT1; M-CPT1; M-cpti; muscle-type carnitine palmitoyltransferase I; zgc:103709
Rabbit
Protein A
RUO
12895, 25756
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.