missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPT1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569410
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 93%; Zebrafish: 92%.
The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and read-through transcripts are expressed from the upstream locus that include exons from this gene.
Specifications
| CPT1B | |
| Polyclonal | |
| Unconjugated | |
| CPT1B | |
| carnitine O-palmitoyltransferase 1, muscle isoform; carnitine O-palmitoyltransferase 1, muscle isoform; synaptonemal complex central element protein 3; carnitine O-palmitoyltransferase 1B; carnitine O-palmitoyltransferase I, mitochondrial muscle isoform; carnitine O-palmitoyltransferase I, muscle isoform; Carnitine palmitoyltransferase 1 beta, muscle isoform; Carnitine palmitoyltransferase 1, muscle; carnitine palmitoyltransferase 1A like; carnitine palmitoyltransferase 1B; carnitine palmitoyltransferase 1B (muscle); carnitine palmitoyltransferase 1B isoform a; carnitine palmitoyltransferase 1b, muscle; carnitine palmitoyltransferase I; carnitine palmitoyltransferase Ibeta 1; carnitine palmitoyltransferase Ibeta 2; carnitine palmitoyltransferase Ibeta 3; carnitine palmitoyltransferase I-like protein; CPT I; Cpt1; cpt1al; CPT1B; CPT1M; Cpt1-m; CPTI; CPTIB; CPT-IB; CPT-Ibeta 1; CPT-Ibeta 3; CPTI-M; EC 2.3.1.21; FLJ55729; FLJ58750; hypothetical protein LOC449677; KIAA1670; LOW QUALITY PROTEIN: carnitine O-palmitoyltransferase 1, muscle isoform; MCCPT1; MCPT1; M-CPT1; M-cpti; muscle-type carnitine palmitoyltransferase I; zgc:103709 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12895, 1375 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q924X2, Q92523 | |
| CPT1B | |
| synthetic peptide directed towards the middle region of human CPT1B. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction