missing translation for 'onlineSavingsMsg'
Learn More

CPT1B Antibody (43), HRP, Novus Biologicals™

Product Code. 18343601 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343601 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343601 Supplier Novus Biologicals Supplier No. NBP290486H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CPT1B Monoclonal specifically detects CPT1B in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPT1B
Applications ELISA
Classification Monoclonal
Clone 43
Conjugate HRP
Formulation PBS
Gene Alias carnitine O-palmitoyltransferase 1, muscle isoform, carnitine O-palmitoyltransferase I, mitochondrial muscle isoform, Carnitine O-palmitoyltransferase I, muscle isoform, Carnitine palmitoyltransferase 1B, carnitine palmitoyltransferase 1B (muscle), Carnitine palmitoyltransferase I-like protein, CPT I, CPT1M, CPT1-MMCCPT1, CPTI, CPTI-M, EC 2.3.1, EC 2.3.1.21, FLJ55729, FLJ58750, KIAA1670, MCPT1, M-CPT1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CPT1B (NP_032596.1; Met 1-Lys 246).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1375
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.