missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPT1A Recombinant Rabbit Monoclonal Antibody (3M6E9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551274
This item is not returnable.
View return policy
Description
Immunogen sequence: WDIPGECQEV IETSLNTANL LANDVDFHSF PFVAFGKGII KKCRTSPDAF VQLALQLAHY KDMGKFCLTY EASMTRLFRE GRTETVRSCT TESCDFVRAM VDPAQTVEQR LKLFKLASEK HQHMYRLAMT GSGIDRHLFC LYVVSKYLAV ESPFLKEVLS EPWRLSTSQT PQQQVELFDL ENNPEYVSSG GGFGPVADDG Y.
The mitochondrial oxidation of long-chain fatty acids is initiated by the sequential action of carnitine palmitoyltransferase I (which is located in the outer membrane and is detergent-labile) and carnitine palmitoyltransferase II (which is located in the inner membrane and is detergent-stable), together with a carnitine-acylcarnitine translocase. CPT I is the key enzyme in the carnitine-dependent transport across the mitochondrial inner membrane and its deficiency results in a decreased rate of fatty acid beta-oxidation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| CPT1A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P50416 | |
| Cpt1a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 520-720 of human CPT1A (NP_001867.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
| ELISA, Western Blot | |
| 3M6E9 | |
| Unconjugated | |
| Cpt1a | |
| C730027G07; carnitine O-palmitoyltransferase; carnitine O-palmitoyltransferase 1, liver isoform; Carnitine O-palmitoyltransferase I, liver isoform; carnitine palmitoyl transferase I isoform; Carnitine palmitoyltransferase 1 alpha, liver isoform; carnitine palmitoyltransferase 1, liver; carnitine palmitoyltransferase 1A; carnitine palmitoyltransferase 1A (liver); carnitine palmitoyltransferase 1a, liver; carnitine palmitoyltransferase I, liver; CPT 1; CPT I; Cpt1; Cpt-1; CPT1 L; CPT1A; CPT1-L; CPTI; CPT-Ia; CPTI-L; L-CPT I; L-CPT1; mitochondrial carnitine palmitoyltransferase 1A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1374 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido