Learn More
CPT1A Mouse anti-Human, CoraLite 647, Clone: 5D8G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL647-66039-100UL
Description
The CPT1A antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human CPT1A. This antibody recognizes human, mouse, rat antigen. The CPT1A antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™647.
Specifications
| CPT1A | |
| Monoclonal | |
| CoraLite 647 | |
| 100 μL | |
| 1374 | |
| IgG2a |
| Flow Cytometry (Intracellular Staining) | |
| 5D8G9 | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |