missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT1A Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33364-100ul
This item is not returnable.
View return policy
Description
CPT1A Monoclonal antibody specifically detects CPT1A in Human samples. It is validated for ELISA,Western Blot
Specifications
| CPT1A | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| carnitine O-palmitoyltransferase 1, liver isoform, Carnitine O-palmitoyltransferase I, liver isoform, Carnitine palmitoyltransferase 1A, carnitine palmitoyltransferase 1A (liver), carnitine palmitoyltransferase I, liver, CPT I, CPT1, CPT1-LEC 2.3.1.21, CPTI-L, EC 2.3.1, L-CPT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 520-720 of human CPT1A (NP_001867.2).,, Sequence:, WDIPGECQEVIETSLNTANLLANDVDFHSFPFVAFGKGIIKKCRTSPDAFVQLALQLAHYKDMGKFCLTYEASMTRLFREGRTETVRSCTTESCDFVRAMVDPAQTVEQRLKLFKLASEKHQHMYRLAMTGSGIDRHLFCLYVVSKYLAVESPFLKEVLSEPWRLSTSQTPQQQVELFDLENNPEYVSSGGGFGPVADDGY | |
| 100 μL | |
| Neuroscience | |
| 1374 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction