missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF73 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-57528UV
This item is not returnable.
View return policy
Description
CPSF73 Polyclonal antibody specifically detects CPSF73 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CPSF73 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| cleavage and polyadenylation specific factor 3, 73kD subunit, cleavage and polyadenylation specific factor 3, 73kDa, cleavage and polyadenylation specificity factor 3, Cleavage and polyadenylation specificity factor 73 kDa subunit, cleavage and polyadenylation specificity factor subunit 3, CPSF 73 kDa subunit, CPSF-73, CPSF73CPSF, EC 3.1.27, EC 3.1.27.-, mRNA 3'-end-processing endonuclease CPSF-73, YSH1 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human cleavage and polyadenylation specific factor 3, 73kDa using the numbering given in entry NP_057291.1 (GeneID 51692). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51692 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction