missing translation for 'onlineSavingsMsg'
Learn More

CPSF73 Antibody (6E6), Novus Biologicals™

Product Code. 18394639 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394639 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394639 Supplier Novus Biologicals Supplier No. H00051692M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CPSF73 Monoclonal antibody specifically detects CPSF73 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF73
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 6E6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100 to 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 to 1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_057291
Gene Alias cleavage and polyadenylation specific factor 3, 73kD subunit, cleavage and polyadenylation specific factor 3, 73kDa, cleavage and polyadenylation specificity factor 3, Cleavage and polyadenylation specificity factor 73 kDa subunit, cleavage and polyadenylation specificity factor subunit 3, CPSF 73 kDa subunit, CPSF-73, CPSF73CPSF, EC 3.1.27, EC 3.1.27.-, mRNA 3'-end-processing endonuclease CPSF-73, YSH1
Host Species Mouse
Immunogen CPSF3 (NP_057291, 585 a.a. ~ 684 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LEVQSNPKIRKGAVQKVSKKLEMHVYSKRLEIMLQDIFGEDCVSVKDDSILSVTVDGKTANLNLETRTVECEEGSEDDESLREMVELAAQRLYEALTPVH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51692
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.