Learn More
Invitrogen™ CPSF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596485
Description
Positive Samples: HeLa, 293T, Jurkat, HepG2, HT-1080, Mouse brain, Mouse liver, Rat brain; Cellular Location: Nucleus Immunogen sequence: MADGVDHIDI YADVGEEFNQ EAEYGGHDQI DLYDDVISPS ANNGDAPEDR DYMDTLPPTV GDDVGKGAAP NVVYTYTGKR IALYIGNLTW WTTDEDLTEA VHSLGVNDIL EIKFFENRAN GQSKGFALVG VGSEASSKKL MDLLPKRELH GQNPVVTPCN KQFLSQFEMQ SRKTTQSGQM SGEGKAGPPG GSSRAAFPQG GRGRGRFPGA VPGGDRFPGP AGPGGPPPPF PAGQTPPRPP LGPPGPPGPP GPPPPGQVLP PPLAGPPNRG DRPPPPVLFP GQPFGQPPLG PLPPGPPPPV PGYGPPPGPP PPQQGPPPPP GPFPPRPPGP.
CPSF6 is one subunit of a cleavage factor required for 3' RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3' end processing complex and facilitates the recruitment of other processing factors. The cleavage factor complex is composed of four polypeptides. The protein encoded by this gene is one subunit of a cleavage factor required for 3' RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3' end processing complex and facilitates the recruitment of other processing factors. The cleavage factor complex is composed of four polypeptides. This gene encodes the 68kD subunit. It has a domain organization reminiscent of spliceosomal proteins.
Specifications
| CPSF6 | |
| Polyclonal | |
| Unconjugated | |
| CPSF6 | |
| 4733401N12Rik; AI256641; CFIM; CFIM68; cleavage and polyadenylation specific factor 6; cleavage and polyadenylation specific factor 6, 68 kD subunit; cleavage and polyadenylation specific factor 6, 68kDa; cleavage and polyadenylation specificity factor 68 kDa subunit; Cleavage and polyadenylation specificity factor subunit 6; cleavage factor Im complex 68 kDa subunit; CPSF 68 kDa subunit; CPSF6; cpsf6 protein; HPBRII-4; HPBRII-7; pre-mRNA cleavage factor I, 68kD subunit; pre-mRNA cleavage factor Im (68kD); pre-mRNA cleavage factor Im 68 kDa subunit; Protein HPBRII-4/7; Unknown (protein for MGC:142454); wu:fa22f12; zgc:85819 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11052, 299811, 432508 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16630, Q6NVF9 | |
| CPSF6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-330 of human CPSF6 (NP_0089382). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.