missing translation for 'onlineSavingsMsg'
Learn More

CPSF6 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18377146 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377146 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377146 Supplier Novus Biologicals Supplier No. NBP299189AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CPSF6 Polyclonal specifically detects CPSF6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF6
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias CFIM, CFIm68, CFIM68CPSF 68 kDa subunit, CFIMcleavage and polyadenylation specific factor 6, 68kD subunit, cleavage and polyadenylation specific factor 6, 68kDa, Cleavage and polyadenylation specificity factor 68 kDa subunit, cleavage and polyadenylation specificity factor subunit 6, HPBRII-4, HPBRII-7, pre-mRNA cleavage factor I, 68kD subunit, pre-mRNA cleavage factor Im (68kD), Pre-mRNA cleavage factor Im 68 kDa subunit, Protein HPBRII-4/7
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human CPSF6.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11052
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.