missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF4 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79826R
This item is not returnable.
View return policy
Description
CPSF4 Polyclonal antibody specifically detects CPSF4 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CPSF4 | |
| Polyclonal | |
| Western Blot | |
| 30kD, cleavage and polyadenylation specific factor 4, 30kD subunit, cleavage and polyadenylation specific factor 4, 30kDa, CPSF 30 kDa subunit, Neb-1, No arches homolog, no arches-like zinc finger protein, NS1 effector domain-binding protein 1 | |
| The immunogen recognized by this antibody maps to a region between residue 220 and 245 of human cleavage and polyadenylation specific factor 4, 30kD subunit using the numbering given in entry NP_006684.1 (GeneID 10898). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 10898 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction