missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPSF3 Recombinant Rabbit Monoclonal Antibody (23GB3105), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | CPSF3 |
|---|---|
| Clone | 23GB3105 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089041
![]() |
Invitrogen™
MA552538 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089733
|
Invitrogen™
MA552539 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CPSF3 Recombinant Monoclonal antibody specifically detects CPSF3 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| CPSF3 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9QXK7, Q9UKF6 | |
| 298916, 51692, 54451 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3105 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| cleavage and polyadenylation specific factor 3; cleavage and polyadenylation specific factor 3, 73kDa; cleavage and polyadenylation specificity factor 3; cleavage and polyadenylation specificity factor 73 kDa subunit; Cleavage and polyadenylation specificity factor subunit 3; CPSF; CPSF 73 kDa subunit; Cpsf3; Cpsf73; CPSF-73; mRNA 3'-end-processing endonuclease CPSF-73; YSH1 | |
| A synthesized peptide derived from CPSF73 (600-684AA). | |
| Primary | |
| -20°C | |
| Cpsf3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title