Learn More
Invitrogen™ CPSF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555023
Description
Immunogen sequence: IELRKRVKLE GKELEEYLEK EKLKKEAAKK LEQSKEADID SSDESDIEED IDQPSAHKTK HDLMMKGEGS RKGSFFKQAK KSYPMFPAPE ERIKWDEYGE IIKPEDFLVP ELQATEEEKS KLESGLTNGD EPMDQDLSDV PTKCISTTES I Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
CPSF2 gene ontology annotations related to this gene include mRNA cleavage; mRNA polyadenylation.
Specifications
| CPSF2 | |
| Polyclonal | |
| Unconjugated | |
| CPSF2 | |
| 100kDa; 2610024B04Rik; AI662483; cleavage and polyadenylation specific factor 2; cleavage and polyadenylation specific factor 2, 100kD subunit; cleavage and polyadenylation specific factor 2, 100kDa; cleavage and polyadenylation specificity factor 100 kDa subunit; Cleavage and polyadenylation specificity factor subunit 2; Cpsf; CPSF 100 kDa subunit; CPSF 100kDa subunit; CPSF100; CPSF2; KIAA1367; Mcpsf; mKIAA1367 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 299256, 51786, 53981 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O35218, Q9P2I0 | |
| CPSF2 | |
| Recombinant protein corresponding to Human CPSF2. Recombinant protein control fragment (Product #RP-88985). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.