missing translation for 'onlineSavingsMsg'
Learn More

CPSF2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18385222
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18385222 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18385222 Supplier Bio-Techne Supplier No. NBP306492AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CPSF2 Polyclonal antibody specifically detects CPSF2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias cleavage and polyadenylation specific factor 2, 100kDa, Cleavage and polyadenylation specificity factor 100 kDa subunit, cleavage and polyadenylation specificity factor subunit 2, CPSF 100 kDa subunit, CPSF 100kDa subunit, CPSF100, KIAA1367cleavage and polyadenylation specific factor 2, 100kD subunit
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CPSF2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53981
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.