missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11031-1-AP-150UL
This item is not returnable.
View return policy
Description
Midline-1 (Tripartite motif-containing protein 18, Putative transcription factor XPRF, RING finger protein 59) is a 667 amino acid protein encoded by the human gene MID1. Midline-1 belongs to the TRIM/RBCC family and contains two B box-type zinc fingers, one B30.2/SPRY domain, one COS domain, one fibronectin type-III domain and one RING-type zinc finger. Midline-1 is believed to have E3 ubiquitin ligase activity which targets the catalytic subunit of protein phosphatase 2 for degradation. It is a cytoplasmic protein found as a homodimer or heterodimer with Midline-2. It also interacts with IGBP1 (Lymphocyte signaling protein A4). Defects in MID1 are the cause of Opitz syndrome type I (OS-I). OS-I is an X-linked recessive disorder characterized by hypertelorism, genital-urinary defects such as hypospadias in males and splayed labia in females, lip-palate-laryngotracheal clefts, imperforate anus, developmental delay and congenital heart defects.Specifications
| CPSF1 | |
| Polyclonal | |
| Unconjugated | |
| CPSF1 | |
| CPSF 160 kDa subunit, CPSF1, CPSF160, HSU37012, P/cl.18 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29894, 366952, 94230 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q10570, Q9EPU4 | |
| Cpsf1 | |
| CPSF1 Fusion Protein Ag1489 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion