missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPSF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566747
This item is not returnable.
View return policy
Description
Immunogen sequence: PHWLLVTGRGA LRLHPMAIDG PVDSFAPFHN VNCPRGFLYF NRQGELRISV LPAYLSYDAP WPVRKIPLRC TAHYVAYHVE SKVYAVATST NTPCARIPRM TGEEKEFETI ERDERYIHPQ QEAFSIQLIS PVSWEAIPNA RIELQEWE Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
Midline-1 (Tripartite motif-containing protein 18, Putative transcription factor XPRF, RING finger protein 59) is a 667 amino acid protein encoded by the human gene MID1. Midline-1 belongs to the TRIM/RBCC family and contains two B box-type zinc fingers, one B30.2/SPRY domain, one COS domain, one fibronectin type-III domain and one RING-type zinc finger. Midline-1 is believed to have E3 ubiquitin ligase activity which targets the catalytic subunit of protein phosphatase 2 for degradation. It is a cytoplasmic protein found as a homodimer or heterodimer with Midline-2. It also interacts with IGBP1 (Lymphocyte signaling protein A4). Defects in MID1 are the cause of Opitz syndrome type I (OS-I). OS-I is an X-linked recessive disorder characterized by hypertelorism, genital-urinary defects such as hypospadias in males and splayed labia in females, lip-palate-laryngotracheal clefts, imperforate anus, developmental delay and congenital heart defects.
Specifications
| CPSF1 | |
| Polyclonal | |
| Unconjugated | |
| Cpsf1 | |
| cleavage and polyadenylation specific factor 1; cleavage and polyadenylation specific factor 1, 160kDa; cleavage and polyadenylation specificity factor 1; cleavage and polyadenylation specificity factor 160 kDa subunit; Cleavage and polyadenylation specificity factor subunit 1; CPSF 160 kDa subunit; CPSF1; CPSF160; HSU37012; P/cl.18; polyadenylation specificity factor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29894 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q10570 | |
| Cpsf1 | |
| Recombinant Human CPSF1. Recombinant protein control fragment (Product #RP-107404). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction