missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35723-100ul
This item is not returnable.
View return policy
Description
CPSF1 Polyclonal antibody specifically detects CPSF1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| CPSF1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| cleavage and polyadenylation specific factor 1, 160kD subunit, cleavage and polyadenylation specific factor 1, 160kDa, Cleavage and polyadenylation specificity factor 160 kDa subunit, cleavage and polyadenylation specificity factor subunit 1, CPSF 160 kDa subunit, CPSF160, HSU37012, P/cl.18, polyadenylation specificity factor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 700-950 of human CPSF1 (NP_037423.2).,, Sequence:, LYRDLSGMFTTESRLGGARDELGGRSGPEAEGLGSETSPTVDDEEEMLYGDSGSLFSPSKEEARRSSQPPADRDPAPFRAEPTHWCLLVRENGTMEIYQLPDWRLVFLVKNFPVGQRVLVDSSFGQPTTQGEARREEATRQGELPLVKEVLLVALGSRQSRPYLLVHVDQELLIYEAFPHDSQLGQGNLKVRFKKVPHNINFREKKPKPSKKKAEGGGAEEGAGARGRVARFRYFEDIYGYSGVFICGPSP | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 29894 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction