missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CPS2

Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
CPS2 is a potent and selective CDK2 Degrader (PROTAC™) (IC50 = 24 nM). It induces CDK2 degradation in several AML cell lines, including NB4, U937, OCI-AML2, OCI-AML3 and Kasumi-1 cells. It induces differentiation of AML cell lines, primary AML cells, and granulocytic differentiation of HSCs.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 4-[[5-Amino-1-[(3-methyl-2-thienyl)carbonyl]-1H-1,2,4-triazol-3-yl]amino]-N-[2-[[1-[2-[2-[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]amino]ethoxy]ethoxy]ethyl]-1H-1,2,3-triazol-4-yl]methoxy]ethyl]benzenesulfonamide |
| CAS | 2756741-90-7 |
| Quantity | 2 mg |
| Target | Active Degraders |
| Molecular Formula | C38H42N12O10S2 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction