missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPS1 Polyclonal Antibody

CPS1 Polyclonal Antibody
Brand: Invitrogen PA576339
This item is not returnable.
View return policy
Description
CPS1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| CPS1 | |
| Polyclonal | |
| Unconjugated | |
| CPS1 | |
| 4732433M03Rik; carbamoyl-phosphate synthase (ammonia); carbamoyl-phosphate synthase [ammonia], mitochondrial; carbamoyl-phosphate synthase 1; carbamoyl-phosphate synthase 1, mitochondrial; carbamoyl-phosphate synthetase 1; carbamoyl-phosphate synthetase 1, mitochondrial; carbamoylphosphate synthetase I; carbamoyl-phosphate synthetase I; carbamyl phosphate synthetase I; carboamyl-phosphate synthetase 1; cps; CPS1; CPSase I; CPSASE1; CPSI; D1Ucla3; PHN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1373, 227231, 497840 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P07756, P31327, Q8C196 | |
| CPS1 | |
| Recombinant Protein of human CPS1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction