missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582760
This item is not returnable.
View return policy
Description
Immunogen sequence: FKIPQKGILI GIQQSFRPRF LGVAEQLHNE GFKLFATEAT SDWLNANNVP ANPVAWPSQE GQNPSLSSIR KLIRDGSIDL VINLPNNNTK FVHDNYVIRR TAVDSGIPLL TNFQVTKLFA EAVQKSRKVD SKSLFHYRQY SA Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 98%.
The mitochondrial enzyme encoded by this gene catalyzes synthesis of carbamoyl phosphate from ammonia and bicarbonate. This reaction is the first committed step of the urea cycle, which is important in the removal of excess urea from cells. The encoded protein may also represent a core mitochondrial nucleoid protein. Three transcript variants encoding different isoforms have been found for this gene. The shortest isoform may not be localized to the mitochondrion. Mutations in this gene have been associated with carbamoyl phosphate synthetase deficiency, susceptibility to persistent pulmonary hypertension, and susceptibility to venoocclusive disease after bone marrow transplantation.
Specifications
| CPS1 | |
| Polyclonal | |
| Unconjugated | |
| CPS1 | |
| 4732433M03Rik; carbamoyl-phosphate synthase (ammonia); carbamoyl-phosphate synthase [ammonia], mitochondrial; carbamoyl-phosphate synthase 1; carbamoyl-phosphate synthase 1, mitochondrial; carbamoyl-phosphate synthetase 1; carbamoyl-phosphate synthetase 1, mitochondrial; carbamoylphosphate synthetase I; carbamoyl-phosphate synthetase I; carbamyl phosphate synthetase I; carboamyl-phosphate synthetase 1; cps; CPS1; CPSase I; CPSASE1; CPSI; D1Ucla3; PHN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1373 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P31327 | |
| CPS1 | |
| Recombinant protein corresponding to Human CPS1. Recombinant protein control fragment (Product #RP-102361). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction