missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPS1 Antibody (S01-2A0), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | CPS1 |
|---|---|
| Clone | S01-2A0 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18657043
|
Novus Biologicals
NBP3-19851-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677903
|
Novus Biologicals
NBP3-19851-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CPS1 Monoclonal antibody specifically detects CPS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CPS1 | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 1373 | |
| IgG | |
| Affinity purified |
| S01-2A0 | |
| Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| carbamoyl-phosphate synthase [ammonia], mitochondrial, carbamoyl-phosphate synthase 1, mitochondrial, carbamoyl-phosphate synthetase 1, mitochondrial, carbamoylphosphate synthetase I, Carbamoyl-phosphate synthetase I, CPSase I, CPSASE1, EC 6.3.4.16 | |
| A synthetic peptide of human CPS1 (Uniprot # P31327) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title