missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPOX Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92779-0.02ml
This item is not returnable.
View return policy
Description
CPOX Polyclonal antibody specifically detects CPOX in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CPOX | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Coprogen oxidase, coproporphyrinogen oxidase, coproporphyrinogenase, coproporphyrinogen-III oxidase, mitochondrial, COX, CPO, CPXcoproporphyrinogen oxidase (coproporphyria, harderoporphyria), EC 1.3.3.3, HCP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 111-454 of human CPOX (NP_000088.3). TSLGRPEEEEDELAHRCSSFMAPPVTDLGELRRRPGDMKTKMELLILETQAQVCQALAQVDGGANFSVDRWERKEGGGGISCVLQDGCVFEKAGVSISVVHGNLSEEAAKQMRSRGKVLKTKDGKLPFCAMGVSSVIHPKNPHAPTIHFNYRYFEVEEADGNKQWWFGGGCDLTPTYLNQEDAVHFHRTLKEACDQHGPDLYPKFKKWCDDYFFIAHRGERRGIGGIFFDDLDSPSKEEVFRFVQSCARAVVPSYIPLVKKHCDDSFTPQEKLWQQLRRGRYVEFNLLYDRGTKFGLFTPGSRIESILMSLPLTARWEYMHSPSENSKEAEILEVLRHPRDWVR | |
| 0.02 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 1371 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction